Learn More
Tris[[2-(tert-butoxycarbonyl)ethoxy]methyl]methylamine, Tris[[2-(tert-butoxycarbonyl)ethoxy]methyl]methylamine, TRC
Product Code. 17952065
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952065 5 g 5g
17951625 1 g 1g
17984315 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17952065 Supplier Toronto Research Chemicals Supplier No. TRCT8762005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C25 H47 N O9
Molecular Weight (g/mol) 505.642
Synonym 3,3'-[[2-Amino-2-[[3-(1,1-dimethylethoxy)-3-oxopropoxy]methyl]-1,3-propanediyl]bis(oxy)]bispropanoic Acid 1,1'-Bis(1,1-dimethylethyl) Ester, 3,3'-[[2-Amino-2-[[3-(1,1-dimethylethoxy)-3-oxopropoxy]methyl]-1,3-propanediyl]bis(oxy)]bispropanoic Acid Bis(1,1-dimethylethyl) Ester
IUPAC Name tert-butyl 3-[2-amino-3-[3-[(2-methylpropan-2-yl)oxy]-3-oxopropoxy]-2-[[3-[(2-methylpropan-2-yl)oxy]-3-oxopropoxy]methyl]propoxy]propanoate
SMILES CC(C)(C)OC(=O)CCOCC(N)(COCCC(=O)OC(C)(C)C)COCCC(=O)OC(C)(C)C
Specifications
CAS 175724-30-8
Quantity 5 g
InChI Formula InChI=1S/C25H47NO9/c1-22(2,3)33-19(27)10-13-30-16-25(26,17-31-14-11-20(28)34-23(4,5)6)18-32-15-12-21(29)35-24(7,8)9/h10-18,26H2,1-9H3
Formula Weight 505.3251
Chemical Name or Material Tris[(2-(tert-butoxycarbonyl)ethoxy)methyl]methylamine