Learn More
Triptophenolide, MedChemExpress
Description
Triptophenolide is a colorless crystalline plate isolated from ethyl acetate extracts of Tripterygium wilfordii.
Specifications
Specifications
| Chemical Name or Material | Triptophenolide |
| CAS | 74285-86-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H24O3 |
| Quantity | 5 mg |
| Synonym | Hypolide (+)-Triptophenolide |
| Solubility Information | DMSO : 100 mg/mL (320.10 mM; Need ultrasonic) |
| SMILES | O=C1C(CC[C@]2(C)C3=C(CC[C@]24[H])C(O)=C(C(C)C)C=C3)=C4CO1 |
| Molecular Weight (g/mol) | 312.4 |
| Show More |