Learn More
Usbeck Laborgeraete™ Tripod Plate
Specifications
Specifications
| Material | Stainless Steel 18/8 |
| Length (Metric) | 300 mm |
| Width (Metric) | 150 mm |
| Quantity | 1 Each |
| Type | Tripod Plate |