missing translation for 'onlineSavingsMsg'
Learn More

Triolein, ≥95%, MP Biomedicals™

Product Code. p-4404560 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 11403521

Brand: MP Biomedicals 0210312201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Triolein is used as a substrate for microbial lipases. Triolein is a symmetrical triglyceride

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-32-7
Molecular Formula C57H104O6
Molecular Weight (g/mol) 885.453
MDL Number MFCD00137563
InChI Key PHYFQTYBJUILEZ-BSCDBXJPSA-N
Synonym triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate
PubChem CID 45253964
IUPAC Name [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC

Specifications

Color Yellow
Density 0.91g/mL(lit.)
Assay Percent Range ≥95%
Quantity 1 g
Formula Weight 885.46
Physical Form Liquid
Chemical Name or Material Triolein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.