missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1225
This item is not returnable.
View return policy
Description
TRIM4 Polyclonal specifically detects TRIM4 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| TRIM4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| RING finger protein 87, RNF87tripartite motif protein 4, tripartite motif containing 4, tripartite motif-containing 4, tripartite motif-containing protein 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported human isoforms (as represented by NP_148977 and NP_149082). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_148977 | |
| TRIM4 | |
| Peptide with sequence C-ASLVIPPVTDRK corresponding to C-Terminus according to NP_148977.1, NP_149082.1. | |
| 0.1 mg | |
| Zinc Finger | |
| 89122 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction