Learn More
Trilaciclib hydrochloride, MedChemExpress
Product Code. 16494616
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494616 100 mg 100mg
16489906 50 mg 50mg
16466107 5 mg 5mg
16403388 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16494616 Supplier Medchem Express Supplier No. HY101467A
MedChemExpress Trilaciclib hydrochloride (G1T28 hydrochloride) is a CDK4/6 inhibitor with IC50s of 1 nM and 4 nM for CDK4 and CDK6, respectively.
Trilaciclib hydrochloride (G1T28 hydrochloride) is a CDK4/6 inhibitor with IC50s of 1 nM and 4 nM for CDK4 and CDK6, respectively.
Specifications
Chemical Name or Material Trilaciclib hydrochloride
CAS 1977495-97-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H32Cl2N8O
Quantity 100 mg
Synonym G1T28 hydrochloride
Solubility Information H2O : 25.64 mg/mL (49.36 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 1.1 mg/mL (2.12 mM; Need ultrasonic)
SMILES O=C1C2=CC3=CN=C(NC4=CC=C(N5CCN(C)CC5)C=N4)N=C3N2C6(CCCCC6)CN1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 519.47
Formula Weight 519.47
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease