missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, Reagent Grade, 98%, Honeywell™

Product Code. p-7122142 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
20 L
Packaging:
Glass Bottle
Stainless steel can
Unit Size:
1 L
20 L
This item is not returnable. View return policy

Product Code. 15688180

Brand: Honeywell Research Chemicals T604531L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -76°C (literature)
Density 0.891 g/mL at 25 °C(lit.)
Boiling Point 146°C (literature)
Packaging Glass Bottle
Linear Formula CH(OC2H5)3
MDL Number MFCD00131528
Quantity 1 L
UN Number UN2524
Beilstein 605384
Vapor Density 5.11 (vs air)
Percent Purity >97.5%
Grade Reagent
Chemical Name or Material Triethyl orthoformate, Reagent grade, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.