missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Tricoton Heavy Gloves
Click to view available options
:
Size: 9
Description
- Good for handling of small parts, possibly up to100°C hot.
- Knitting thickness 7-heavy.
- Can be worn as an under-glove.
- Heavy knit.
- Close fitting.
- Good sense of touch.
- Comfortable.
- Neutral pH.
- Sweat absorbing.
- Shock absorbing.
- 100% cotton (untreated).
- Elasticized cuff.
- Brightly colored to promote hygiene and cleanliness.
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY II Medium risks, EN 420-94 EEC General requirements, EN 388-94 Mechanical hazards, EN 407-94 Heat and/or fire, EC category PSA 2, Quality assurance ISO 9001/2000, EC certification number N ° 0075/797/162/05/08/0165, EC-type-examination Certificate N ° 0075/797/162/05/08/0165. |
| Classification | Touch: 5 |
| Construction | String Knit |
| Cut Resistance Rating | 1 |
| Material | Cotton |
| Product Type | Gloves |
| Size | 9 |
| For Use With (Application) | Gastronomy, Food, Logistics, General and Automotive industries and suppliers. |
| Glove Cuff Style | Elasticated |
| Quantity | 600/PK |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction