missing translation for 'onlineSavingsMsg'
Learn More

Tributyltin Chloride, Tributyltin Chloride, TRC

Product Code. 30138045
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138045 5 g 5g
30041295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138045 Supplier Toronto Research Chemicals Supplier No. TRCT7738105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12 H27 Cl Sn
Molecular Weight (g/mol) 325.51
Synonym TBTC, Tributyltin chloride
IUPAC Name tributyl(chloro)stannane
SMILES CCCC[Sn](Cl)(CCCC)CCCC

Specifications

CAS 1461-22-9
Quantity 5 g
InChI Formula InChI=1S/3C4H9.ClH.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1H;/q;;;;+1/p-1
Formula Weight 326.0823
Chemical Name or Material Tributyltin Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.