Learn More
Triamcinolone acetonide, MedChemExpress
Product Code. 16472249
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16472249
Brand: Medchem Express HYB0636
MedChemExpress Triamcinolone acetonide is a more potent type of triamcinolone, being about 8 times as effective as prednisone.
Triamcinolone acetonide is a more potent type of triamcinolone, being about 8 times as effective as prednisone.
Specifications
Chemical Name or Material Triamcinolone acetonide
CAS 76-25-5
Color White
Purity Grade Notes Research
Molecular Formula C24H31FO6
Quantity 1 g
Solubility Information DMSO : 50 mg/mL (115.07 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.23 mM; Need ultrasonic)
SMILES OCC([C@]12[C@@]3([C@@]([C@@]4([H])[C@]([C@H](C3)O)([C@@]5(C(CC4)=CC(C=C5)=O)C)F)([H])C[C@@]1([H])OC(C)(O2)C)C)=O
Molecular Weight (g/mol) 434.5
Formula Weight 434.5
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation