missing translation for 'onlineSavingsMsg'
Learn More

Tri(2-methoxyethoxy)vinylsilane, Dr. Ehrenstorfer

Product Code. 30316294
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316294 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316294 Supplier Dr. Ehrenstorfer Supplier No. DRECA17876450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinyltris(2-methoxyethoxy)silane
CAS 1067-53-4
Analyte or Component Names Vinyltris(2-methoxyethoxy)silane
Shipping Condition +20°C
Molecular Formula C11 H24 O6 Si
Certifications/Compliance ISO/IEC 17025
Synonym Silane, tris(2-methoxyethoxy)vinyl- (6CI,7CI,8CI), 6-Ethenyl-6-(2-methoxyethoxy)-2,5,7,10-tetraoxa-6-silaundecane, A 172, A 172 (coupling agent), A 172NT, A 173, AC 77, CG 172, Crosile 172, DB 172, DB 172 (coupling agent), Dynasylan VTMOEO, FD 172, Fibnasil, GF 58, Geniosil GF 58, KBC 1003, KH 172, KH-A 172, NSC 78465, NUCA 172, SCA 972, SCA-A 172, SCA-V 71C, SG-SI 172, SH 6082, SZ 6082, Silane A 172, Silox VS 924, Silquest A 172, Silquest A 172NT, Struktol SCA 972, Tri(2-methoxyethoxy)vinylsilane, Tris(2-methoxyethoxy)vinylsilane, Tris(methoxyethoxy)vinylsilane, Tris(β-methoxyethoxy)vinylsilane, U 613, UNC-A 172, V 5000, VTMES-SCA 972, VTMOEO, VTS-ME, Vinyltri(methoxyethoxy)silane, Vinyltri(β-methoxyethoxy)silane, Vinyltris(β-methoxyethoxy)silane, WD 27, WD 72, YGO 1204, Z 6082, Z 6172, 2,5,7,10-Tetraoxa-6-silaundecane, 6-ethenyl-6-(2-methoxyethoxy)-
InChI Formula InChI=1S/C11H24O6Si/c1-5-18(15-9-6-12-2,16-10-7-13-3)17-11-8-14-4/h5H,1,6-11H2,2-4H3
SMILES COCCO[Si](OCCOC)(OCCOC)C=C
IUPAC Name ethenyl-tris(2-methoxyethoxy)silane
Molecular Weight (g/mol) 280.39
Quantity 100 mg
Formula Weight 280.1342 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.