missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone 17beta-Glucuronide Sodium Salt, TRC

Product Code. 30452689
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452689 1 mg 1mg
30452047 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452689 Supplier Toronto Research Chemicals Supplier No. TRCT7189951MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trenbolone 17beta-Glucuronide Sodium Salt
Analyte or Component Names Trenbolone 17beta-Glucuronide Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C24 H30 O8
Synonym Trenbolone 17β-Glucuronide Sodium Salt, Trenbolone Glucuronide Sodium Salt, (17β)-3-Oxoestra-4,9,11-trien-17-yl β-D-Glucopyranosiduronic Acid Sodium Salt
InChI Formula InChI=1S/C24H30O8/c1-24-9-8-14-13-5-3-12(25)10-11(13)2-4-15(14)16(24)6-7-17(24)31-23-20(28)18(26)19(27)21(32-23)22(29)30/h8-10,15-21,23,26-28H,2-7H2,1H3,(H,29,30)/t15-,16+,17+,18+,19+,20-,21+,23-,24+/m1/s1
Solution Type Neat
SMILES C[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@H]2O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[[(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl]oxy]tetrahydropyran-2-carboxylic acid
Industry Type Forensics
Molecular Weight (g/mol) 446.49
Quantity 1 mg
Formula Weight 446.1941
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.