missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trenbolone 17beta-Glucuronide Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Trenbolone 17beta-Glucuronide Sodium Salt |
| Analyte or Component Names | Trenbolone 17beta-Glucuronide Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H30 O8 |
| Synonym | Trenbolone 17β-Glucuronide Sodium Salt, Trenbolone Glucuronide Sodium Salt, (17β)-3-Oxoestra-4,9,11-trien-17-yl β-D-Glucopyranosiduronic Acid Sodium Salt |
| InChI Formula | InChI=1S/C24H30O8/c1-24-9-8-14-13-5-3-12(25)10-11(13)2-4-15(14)16(24)6-7-17(24)31-23-20(28)18(26)19(27)21(32-23)22(29)30/h8-10,15-21,23,26-28H,2-7H2,1H3,(H,29,30)/t15-,16+,17+,18+,19+,20-,21+,23-,24+/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@H]2O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[[(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl]oxy]tetrahydropyran-2-carboxylic acid |
| Industry Type | Forensics |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?