missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trastuzumab Antibody (2261E) [Biotin], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB95471B
This item is not returnable.
View return policy
Description
Trastuzumab Monoclonal antibody specifically detects Trastuzumab in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Trastuzumab | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Full-length Trastuzumab | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| 2261E | |
| Western Blot, Flow Cytometry, ELISA | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu