missing translation for 'onlineSavingsMsg'
Learn More

TRAPPC9/NIBP Mouse anti-Human, Mouse, Clone: 1C4F9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66131-1-IG-150UL

 View more versions of this product

Product Code. 16887725

  • 410.00 EUR / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NIBP is a 1246 amino acid intracellular regulatory protein. Reports suggest that NIBP physically interacts with NIK, IKK (beta), but not IKK (alpha) or IKK (gamma) and thus acts as a novel enhancer of cytokine-induced NF-(kappa) B signaling pathway. Due to its neuronal expression NIBP is considered as a potential target for modulating the NF-(kappa) B signaling cascade in neuronal pathologies dependent upon abnormal activation of this pathway. NIBP is mainly expressed in brain, muscle, heart, and kidney.
Specifications

Specifications

TRAPPC9/NIBP
Monoclonal
1.8 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3U0M1, Q96Q05
Trappc9
TRAPPC9,NIBP Fusion Protein Ag8949
150 μL
Primary
Human, Mouse
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
1C4F9
Unconjugated
TRAPPC9
IBP, IKBKBBP, KIAA1882, NIBP, NIK and IKBKB binding protein, T1, TRAPPC9, TRS120, Tularik gene 1 protein
Mouse
Protein A
RUO
76510, 83696
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.