missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRAPPC5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15918084
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15918084 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15918084 Supplier Invitrogen™ Supplier No. PA565326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AVWKALFGKEA DKLEQANDDA RTFYIIEREP LINTYISVPK ENSTLNCASF TAGIVEAVLT HSGFPAKVTA HWHKGTTLMI KFEEAVIARD R Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

TRAPPC5 (trafficking protein particle complex 5), also known as TRS31, is a 181 amino acid protein belonging to the TRAPP (transport protein particle) small subunits family and the BET3 subfamily. Encoded by a gene that maps to human chromosome 19p13.2, TRAPPC5 is part of the multisubunit TRAPP tethering complex, which acts as a GTP exchange factor. Evolutionarily conserved, TRAPPC5 plays a role in protein binding,vesicle-mediated transport and nucleotide exchange stimulation. TRAPPC5 also performs guanine nucleotide exchanger factor (GEF) functions both in vitro and in vivo. Localizing to the Golgi apparatus, TRAPPC5 is essential for endoplasmic reticulum (ER)-to-Golgi and intra-Golgi vesicle trafficking in yeast, as well as additional transport events in mammals, such as post-Golgi trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRAPPC5
Gene Accession No. Q8IUR0
Gene Alias 4021401A16Rik; RGD1563971; trafficking protein particle complex 5; trafficking protein particle complex subunit 5; Trappc5; TRS31
Gene Symbols TRAPPC5
Host Species Rabbit
Immunogen Recombinant Human TRAPPC5. Recombinant protein control fragment (Product #RP-99215).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126003
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.