Learn More
Fisherbrand™ Transparent Terylene Mesh PVC Tubing
Shop All Fisherbrand Products
Click to view available options
Description:
6.5 x 3mm
5 x 3mm
8 x 3mm
10 x 3mm
Unit Size:
10m
Description
All dimensions are approximate.
- Transparent, colorless
- Terylene mesh reinforced wall
- Flexible yet great inherent strength
- Suitable for moderate pressure or vacuum applications
Specifications
Specifications
| Description | 5 x 3mm |
| Material | PVC |
| Overall Length (Metric) | 10 m |
| For Use With (Application) | Suitable For Moderate Pressure Or Vacuum Applications |
| Packaging Quantity | 1 Pack |
| Color | Transparent |