missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Transmissible Gastroenteritis Virus Monoclonal Antibody (TGEV11-314.6), Invitrogen™

Product Code. 17800156
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17800156 100 μg 100µg
11593393 500 μg 1mg
17830276 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17800156 Supplier Invitrogen™ Supplier No. MA546182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10784 detects Transmissible Gastroenteritis Virus. MA1-10784 has been successfully used in ELISA applications. The MA1-10784 immunogen is TGEV-infected ST cells.

Transmissible gastroenteritis virus or Transmissible gastroenteritis coronavirus is a coronavirus which infects pigs. It is an enveloped, positive-sense, single-stranded RNA virus which enters its host cell by binding to the APN receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transmissible Gastroenteritis Virus
Applications ELISA
Classification Monoclonal
Clone TGEV11-314.6
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias TGV
Host Species Mouse
Immunogen TGEV-infected ST cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.