Learn More
Translucent Crucibles
Product Code. 12897723
Click to view available options
Diameter (Metric) Outer:
29mm
38mm
43mm
57mm
67mm
82mm
Unit Size:
Each
Product Code. 12897723
Brand: Yorlab 10444/19
Only null left
Specifications
Color Translucent
Material Silica
Capacity (Metric) 50 mL
Diameter (Metric) Outer 57mm
Height (Metric) 37mm
Product Type Crucible