missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human Caspase-8 siRNA

Product Code. 16069231
Click to view available options
Quantity:
10 nmol
2 nmol
Unit Size:
10nmol
2nmol
This item is not returnable. View return policy

Product Code. 16069231

Brand: enQuireBio™ QX4310nmol

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Blocks transcription and translation of Caspase-8 from Human.

Specifications

Accession Number NP_001219
Cell Lines Tested 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs.
Content And Storage Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Gene Symbol CASP8
Gene ID (Entrez) 841
Product Type siRNA
Quantity 10 nmol
Synonym CAP4, MCH5, FLICE, MACH, Casp-8, ALPS2B, CASP8
Target Human
Form Lyophilized
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.