missing translation for 'onlineSavingsMsg'
Learn More

(-)-trans-Caryophyllene, MP Biomedicals™

Product Code. 11449091 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
Unit Size:
10mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449091 10 mL 10mL
11439091 5 mL 5mL
11459091 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449091 Supplier MP Biomedicals Supplier No. 0215494410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 87-44-5
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
InChI Key NPNUFJAVOOONJE-IZZDOVSWSA-N
Synonym --caryophyllene, caryophyllene, l-caryophyllene, trans-caryophyllene, e-.beta.-caryophyllene, .beta.-caryophyllene,-, caryophyllene ,alpha + beta mixt., + e-caryophyllene, e-.beta.-caryophylene, 8-methylene-4,11-trimethyl bicyclo 7.2.0 undec-4-ene
PubChem CID 5354499
IUPAC Name (4E)-4,11,11-trimethyl-8-methylidenebicyclo[7.2.0]undec-4-ene
SMILES CC1=CCCC(=C)C2CC(C2CC1)(C)C

Specifications

Color Yellow
Density 0.9g/mL
Quantity 10 mL
Formula Weight 204.4
Physical Form Liquid
Chemical Name or Material (-)-Trans-Caryophyllene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.