Learn More
trans-2,cis-6-Nonadienal, 90+% (major isomer), remainder mainly trans,trans-isomer
Product Code. p-7024128
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11473917 1 g 1g
11483917 5 g 5g
11493917 25 g 25g
3 options
Product Code. 11473917
Brand: Thermo Scientific Alfa Aesar A18356.03
Request Bulk or Custom Format
CAS: 557-48-2 | C9H14O | 138.21 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 557-48-2
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00007009
InChI Key HZYHMHHBBBSGHB-ODYTWBPASA-N
Synonym trans-2,cis-6-nonadienal, violet leaf aldehyde, cucumber aldehyde, 2e,6z-nona-2,6-dienal, 2,6-nonadienal, 2e,6z, nona-2,6-dienal, e,z-2,6-nonadienal, 2e,6z-nonadienal, 2e,6z-nona-2,6-dien-1-al, 2-trans-6-cis-nonadienal
PubChem CID 643731
ChEBI CHEBI:7610
IUPAC Name (2E,6Z)-nona-2,6-dienal
SMILES CCC=CCCC=CC=O
Specifications
Density 0.866
Boiling Point 90°C (4 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.474
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 138.2
Percent Purity ≥90%
Assay remainder mainly trans,trans-isomer
Chemical Name or Material trans-2,cis-6-Nonadienal