missing translation for 'onlineSavingsMsg'
Learn More

Tranilast, MedChemExpress

Product Code. p-200099798
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16438408

Brand: Medchem Express HYB0195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tranilast (MK-341) acts as an anti-atopic agent. Tranilast suppresses production of prostaglandin D2 (PGD2, IC50= 0.1 mM). Tranilast sodium exhibits anti-inflammatory and immunomodulatory effects. Tranilast sodium antagonizes angiotensin II and inhibits its biological effects in vascular smooth muscle cells.

Tranilast (MK-341) acts as an anti-atopic agent. Tranilast suppresses production of prostaglandin D2 (PGD2, IC50= 0.1 mM). Tranilast sodium exhibits anti-inflammatory and immunomodulatory effects. Tranilast sodium antagonizes angiotensin II and inhibits its biological effects in vascular smooth muscle cells.

Specifications

Chemical Name or Material Tranilast
CAS 53902-12-8
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C18H17NO5
Quantity 10 mg
Synonym MK-341 SB 252218
Solubility Information DMSO : 50 mg/mL (152.75 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=CC=C1NC(/C=C/C2=CC=C(OC)C(OC)=C2)=O
Molecular Weight (g/mol) 327.33
Formula Weight 327.33
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.