Learn More
Abnova™ TRAF3/CEN14q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FG0218.100uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
CAP-1,CD40bp,CRAF1,LAP1 | |
7187 | |
100 μL |
Tube | |
TNF receptor-associated factor 3 | |
TRAF3 | |
DNA/RNA | |
Liquid |