missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TRAF3/CEN14q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00 EUR - 2430.00 EUR
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | TNF receptor-associated factor 3 |
Gene Alias | CAP-1,CD40bp,CRAF1,LAP1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033355
|
Abnova™
FG0218.100UL |
100 μL |
1475.00 EUR
100µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043355
|
Abnova™
FG0218.200UL |
200 μL |
2430.00 EUR
200µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
CAP-1,CD40bp,CRAF1,LAP1 | |
7187 | |
Liquid |
Tube | |
TNF receptor-associated factor 3 | |
TRAF3 | |
DNA/RNA |