Learn More
TRAF-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NB100-56436SS
 View more versions of this product
Product Code. 18066354
  • 213.00 EUR / 25µg
Description
TRAF-6 Polyclonal specifically detects TRAF-6 in Human samples. It is validated for Western Blot, Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRAF-6
Polyclonal
Unconjugated
0.1 ml PBS with 0.02% Sodium Azide
E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6
Rabbit
Protein G purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry
1.0 mg/mL
Western Blot 3-5 ug/ml, Flow Cytometry
Q9Y4K3
TRAF6
Anti-TRAF6 polyclonal antibody was raised against a peptide corresponding to amino acids between 410 and 460 of human TRAF6.
0.025 mg
Immunology, Innate Immunity, Signal Transduction, Zinc Finger
7189
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions