Learn More
Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Alarm Types:
Preprogrammed -20°C to -15°C
Preprogrammed 2°C to 8°C
Data Memory:
10 Day, 1 min. Interval
30 Day, 3 min. Interval
60 Day, 6 min. Interval
90 Days, 10 min. Interval
missing translation for 'unitSize'
Pack of 40
Description
- Logger generates PDF report
- Download directly to PC
- No Software required
- Compact
- Single-use
- Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
- Press start to begin recording crucial temperature data, 30 minute time delay before logging begins
Specifications
Specifications
| Type | Single-use Datalogger |
| Data Memory | 30 Day, 3 min. Interval |
| Calibration | Calibrated |
| Alarm Types | Preprogrammed -20°C to -15°C |
| Software Type | No software required, download data as PDF report |
| Description | 40/Pk |
| Includes | Batch-calibrated Traceable certificate |
| IP Rating | IP67 |
| For Use With (Application) | Refrigerated transportation samples |
| Accuracy (Temperature) | ±0.5°C from -20° to 40°C, otherwise ±1°C |
| Show More |