Learn More
Fisherbrand™ Traceable™ Hi-Accuracy 0.01° Refrigerator Thermometer
For monitoring temperatures in refrigerator and freezer simultaneously or two refrigerator locations. Fisherbrand Traceable Hi-Accuracy Refrigerator Thermometer permits user one-button self-calibration which reads to 0.01° while simultaneously monitoring temperatures in refrigerator and freezer.
Specifications
| Certifications/Compliance | ISO 17025, A2LA, NIST |
|---|---|
| Diameter (English) Probe | 0.19 in. |
| Dimensions (L x W x H) | 4.5 x 2.75 x 0.75 in. (11.4 x 6.9 x 1.9 cm) |
| For Use With (Application) | Monitoring temperature in refrigerator and freezer simultaneously or two refrigerator locations |
| Height (English) | 0.75 in. |
Description
Triple display simultaneously shows minimum/maximum and current temperatures where minimum/maximum monitors readings overnight, on weekends or for any time period, and displays the exact time and date.
- Temperature-buffered bottle sensor
- Two visual (LEDs) and two audio alerts when temperature rises above or falls below high/low set points
- Programmable alarms in 0.10° increments
- Miniature bottle sealing: 1 × 2.5 in. (2.5 × 6.3cm) filled with nontoxic glycol
- High-impact, chemical-resistant ABS plastic case
- Stand, Velcro™, magnetic strips, and wall mount allow placement of thermometer anywhere
- Used in water baths, heating blocks and incubators
Specifications
- Range: -58 to +158°F and -50 to +70°C
- Resolution: 0.01°
- Accuracy: ±0.3°C
- Weight: 5 oz. (141g)
Certifications
Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
| ISO 17025, A2LA, NIST | |
| 4.5 x 2.75 x 0.75 in. (11.4 x 6.9 x 1.9 cm) | |
| 0.75 in. | |
| 120 in. | |
| 114 mm | |
| 1.9 cm | |
| 0.01°C | |
| 140 g | |
| 4.5 in. | |
| 70°C | |
| −50°C | |
| -50°C to +70°C | |
| 6.9 cm |
| 0.19 in. | |
| Monitoring temperature in refrigerator and freezer simultaneously or two refrigerator locations | |
| 1.9 cm | |
| 0.7 in. | |
| 304.8 cm | |
| ABS Plastic | |
| 5 oz. | |
| 2.75 in. | |
| 158°F | |
| −58°F | |
| -58°F to +158°F | |
| LED |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.