Learn More
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem and 0.25 in. LCD Screen
Product Code. 11759725 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
-50°C to +280°C
-50°C to +300°C
Temperature Range (English):
-58°F to +302°F
-58°F to +536°F
-58°F to +572°F
Accuracy:
±0.4°C
±1°C
Model:
Food/Piercing/Splash-Proof/Drop-Proof Thermometer
Key-Chain Thermometer
Shockproof/Water-resistant Thermometer
Ultra Food/Waterproof/Piercing/Drop-proof Thermometer
Ultra Shockproof/Water-resistant Thermometer
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Temperature Range (English) Accuracy Model unitSize
11759725 -50°C to +300°C -58°F to +572°F ±0.4°C Ultra Food/Waterproof/Piercing/Drop-proof Thermometer Each
11719725 -50°C to +150°C -58°F to +302°F ±1°C Key-Chain Thermometer Each
11785853 -50°C to +300°C -58°F to +572°F ±1°C Shockproof/Water-resistant Thermometer Each
11769725 -50°C to +280°C -58°F to +536°F ±1°C Food/Piercing/Splash-Proof/Drop-Proof Thermometer Each
11799715 -50°C to +300°C -58°F to +572°F ±0.4°C Ultra Shockproof/Water-resistant Thermometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11759725
Traceable™ Digital Thermometer used to measure temperature of gases, food, and semisolids.
  • Short-stem piercing models are ideal for carrying in a pocket
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
  • Traceable to standards provided by NIST
  • Waterproof thermometers (see Models 14-648-43, -44) are ideal for wet labs, wash-down areas and field use
  • 0.375 in. LCD screen thermometers available (see Cat. No.14-648-47); also available with protective guards (see Cat. Nos. 06-664-32 and 06-664-30)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Certifications/Compliance ISO 17025, A2LA, NIST
Depth (English) 1 in.
Depth (Metric) 2.5 cm
Diameter (English) Probe 0.19 in.
For Use With (Application) Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
Includes Battery, Stainless steel probe and Traceable certificate
Length (English) Probe 8 in.
Length (Metric) 240 mm
Length (Metric) Probe 20 cm
Material Stainless Steel
Model Ultra Food/Waterproof/Piercing/Drop-proof Thermometer
Resolution 0.1°C
Weight (English) 1.25 oz.
Weight (Metric) 35 g
Width (English) 3.75 in.
Product Type Digital Thermometer
Length (English) 9.5 in.
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +572°F
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
Accuracy ±0.4°C
Display Digital LCD
Width (Metric) 9.5 cm