Learn More
Thermo Scientific™ TRACE™ GC Columns for Drugs of Abuse
Product Code. 10477514 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
0.2 mm
0.25 mm
Unit Size:
Each
Product Code. 10477514
Brand: Thermo Scientific™ 26AF130P
Only null left
Add to basket
Add to basket
Analyze common drugs of abuse with these GC columns and achieve high performance, sensitivity and reproducibility.
Analyze amphetamines, codeine, morphine, and THC with high performance and reproducibility using Thermo Scientific™ TRACE™ GC Columns for Drugs of Abuse.
Tailored Specifically for Drugs of Abuse:
  • TRACE TR-DoA 5MS: Widely used for the analysis and determination of a range of toxicological target compounds including amphetamines, codeine, and morphine
  • TRACE TR-DoA 35MS: Ideal for the determination of carboxy-THC because of its higher polarity, making it the recommended column for use in drug testing labs for the confirmation of THC
Excellent Quality and Reproducibility:
  • Low bleed even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
TRUSTED_SUSTAINABILITY
Specifications
Max. Temperature 360/370°C
Diameter (Metric) 0.25 mm
Diameter (Metric) Inner 0.25 mm
Product Line TRACE
Stationary Phase Trace™ TR-DoA 35 MS
USP Type G42
For Use With (Application) GC/MS for Amphetamines, Codeine, and Morphine
Film Thickness 0.25 μm
Length (Metric) 15 m