missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRA-2-54 Mouse, anti-Human, Unconjugated, Clone: TRA-2-54/2J, STEMCELL Technologies
Description
- The TRA-2-54/2J antibody reacts with the tissue non-specific isoform of alkaline phosphatase (AP), which is expressed at high levels in undifferentiated pluripotent stem cells such as human and mouse embryonic stem (ES) cells, as well as induced pluripotent stem (iPS) and embryonal carcinoma (EC) cells
- This isozyme of AP is the same as that expressed by liver, bone and kidney cells
- Levels of AP decrease upon differentiation and, thus, the TRA-2-54/2J antibody can be used to monitor the differentiation status of pluripotent stem cells
- The TRA-2-54/2J antibody does not cross-react with other isozymes of AP, which differ through post-translational modification, and reportedly does not recognize AP from cow, dog, goat, Guinea pig, hamster, mouse, rat or sheep, or Old World Monkeys such as the rhesus monkey, baboon and African green monkey
Specifications
Specifications
| Antigen | TRA-2-54 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | TRA-2-54/2J |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Target Species | Non-human Primate, Feline, Human, Pig, Rabbit |
| Isotype | IgG1 κ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?