Learn More
TRA-1-60 (Podocalyxin) Mouse, anti-Human, PE, Clone: TRA-1-60R, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60064PE
Encompass_Preferred
 View more versions of this product
Product Code. 17158801
  • 450.90 EUR / 100 tests
Description
  • The TRA‐1‐60R antibody reacts with TRA-1-60 (Podocalyxin), a > 200 kDa pluripotent stem cell-specific protein expressed on the surface of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells, embryonal carcinoma (EC) cells and embryonic germ (EG) cells, as well as rhesus monkey ES cell lines
  • The epitope contains sialic acid and is associated with a large-molecular-mass transmembrane protein named podocalyxin
  • Though sialylated, the epitope recognized by the TRA‐1‐60R antibody is resistant to treatment with neuraminidase
  • This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850), and TeSR™2 (Catalog #05860)
Specifications
Specifications
TRA-1-60 (Podocalyxin)
Monoclonal
PE
100 Tests
Rhesus Monkey, Human, Rabbit
Pluripotent Stem Cells
Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
TRA-1-60R
Mouse
Primary
IgM κ