missing translation for 'onlineSavingsMsg'
Learn More

TPOP146, MedChemExpress

Product Code. 16484416
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484416 100 mg 100mg
16475857 5 mg 5mg
16443128 10 mg 10mg
16409686 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484416 Supplier Medchem Express Supplier No. HY100697

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TPOP146 is a selective CBP/P300 benzoxazepine bromodomain inhibitor with Kd values of 134 nM and 5.02 μM for CBP and BRD4.

TPOP146 is a selective CBP/P300 benzoxazepine bromodomain inhibitor with Kd values of 134 nM and 5.02 μM for CBP and BRD4.

Specifications

Chemical Name or Material TPOP146
CAS 2018300-62-2
Color White
Purity Grade Notes Research
Molecular Formula C27H35N3O5
Quantity 100 mg
Solubility Information DMSO : 21.43 mg/mL (44.50 mM; ultrasonic and warming and heat to 60°C)
SMILES COC1=CC(OC)=CC(C2=CC(C(N[C@H]3CCCN(C)C3)=O)=C(OCCN(C(CC)=O)C4)C4=C2)=C1
Molecular Weight (g/mol) 481.58
Formula Weight 481.58
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.