Learn More
Ebro Electronic™ TPC 300 Probe
Calibrated replacement probe for EBI 300 TE
Brand: Ebro Electronic™ 1341-6331A
Additional Details : Weight : 0.10000kg
Description
Fast, flexible core temperature measurementsOrder Info
TemperatureRangeMetric
Specifications
Probe | |
Factory-calibrated | |
TPC 300 | |
-35°C to +75°C |
No Display | |
None | |
±0,5°C (-20°C…+40°C) ±0,8°C for the remaining measurement range |