Description
- Large display with intuitive operation
- Actual and set values of key process parameters are shown on the display at the same time
- Also possible to perform graphical time-based evaluation of process parameters
Specifications
Specifications
| Description | Touchscreen controller - Large display panel with intuitive operation, actual and set values of main process parameters are shown simultaneously on the display. In addition, the graphic chronological evaluation of process parameters is displayed. |
| For Use With (Equipment) | BINDER Chambers Series VD |
| Type | Controller |
| Voltage | 120/230 V |