Learn More
Amarell™ Total Immersion Low Temperature Laboratory Thermometer
Click to view available options
Temperature Range (Metric):
-100°C to +30°C
Length (Metric):
350 mm
400 mm
Unit Size:
Each
Description
- Low temperature precision thermometer
- Total immersion
- Richter-top finish with button
- Suitable for government verification
Specifications
Specifications
| Bottle Fill | Toluene |
| For Use With (Application) | Government testing |
| Immersion Type | Total Immersion |
| Length (Metric) | 400 mm |
| Product Type | Low Temperature Precision Thermometer |
| Max. Temperature (Metric) | 30°C |
| Min. Temperature (Metric) | -100°C |
| Temperature Range (Metric) | -100°C to +30°C |
| Calibrated | Yes |
| Subdivision | 5°C |