Learn More
Amarell™ Total Immersion Low Temperature Laboratory Thermometer
Product Code. 10733523
Click to view available options
Temperature Range (Metric):
-100°C to +30°C
Length (Metric):
350 mm
400 mm
Unit Size:
Each
Product Code. 10733523
Brand: Amarell™ L33040
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Precision thermometer, low temperatures, enclosed scale, capillary lens front, total immersion, Richter-top finish with button, suitable for government verification
  • Low temperature precision thermometer
  • Total immersion
  • Richter-top finish with button
  • Suitable for government verification
Specifications
Bottle Fill Toluene
For Use With (Application) Government testing
Immersion Type Total Immersion
Length (Metric) 400 mm
Product Type Low Temperature Precision Thermometer
Max. Temperature (Metric) 30°C
Min. Temperature (Metric) -100°C
Temperature Range (Metric) -100°C to +30°C
Calibrated Yes
Subdivision 5°C