missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Hardness Reagent Sets, For Colorimetric Water Testing

Product Code. 10060921
Click to view available options
Range:
0.00 to 2.70 mg/L
0.00 to 500 mg/L
200 to 500 mg/L as CaCO3
No. of Tests:
100
For Use With (Application):
Calcium Hardness
Total Hardness Medium Range
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10060921

Brand: Hanna Instruments™ HI9373501

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Hardness Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.

These high quality reagents are designed to follow accepted test methods for colorimetric determination of various parameters of water hardness. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.

  • Reagent sets are available for determination of:
    • Calcium Hardness (using liquid or powdered reagents)
    • Calcium and Magnesium Hardness (high range)
    • Magnesium and Total Hardness
    • Total Hardness (low, medium and/or high range)
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability

Specifications

For Use With (Application) Total Hardness Medium Range
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes 2 dropper bottles and 100 individual packets
No. of Tests 100
Quantity 1 Set
Range 200 to 500 mg/L as CaCO3
Test Method EPA recommended method 130.1
Type Water Testing Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Reagents for 100 tests total hardness
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : 2-Amino-2-methyl-1-propanol, EDTA, Tetrasodium TetrahydrateSalt, Propionic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.