missing translation for 'onlineSavingsMsg'
Learn More

Torsemide, MedChemExpress

Product Code. 16418438
Encompass_Preferred
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
This item is not returnable. View return policy

Product Code. 16418438

Brand: Medchem Express HYB0247

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Torsemide (Torasemide) is an orally active loop diuretic. Torsemide has anti-aldosterone and vasodilatory effects. Torsemide also can be used for the research of heart failure, renal disease and hepatic cirrhosis.

Torsemide (Torasemide) is an orally active loop diuretic. Torsemide has anti-aldosterone and vasodilatory effects. Torsemide also can be used for the research of heart failure, renal disease and hepatic cirrhosis.

Specifications

Chemical Name or Material Torsemide
CAS 56211-40-6
Color White
Purity Grade Notes Research
Molecular Formula C16H20N4O3S
Quantity 10 mg
Synonym Torasemide
Solubility Information DMSO : ≥ 3.5 mg/mL (10.05 mM)
SMILES O=S(C1=C(NC2=CC=CC(C)=C2)C=CN=C1)(NC(NC(C)C)=O)=O
Molecular Weight (g/mol) 348.42
Formula Weight 348.42
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.