Learn More
Tork Centrefeed Dispenser M2
Shop All Tork Products
Click to view available options
Model:
Elevation
Performance
Unit Size:
Each
Description
- Improve overall hygiene due to easy cleaning
- Versatile lock mechanism helps control misuse
- One hand operation makes it easy to use
- Free flow dispensing for flexibility and cross contamination control
Specifications
Specifications
| Product Type | Centerfeed Wiper Dispenser |
| Color | Black |
| Alternate Part Number | 7322540349399 |
| Length (Metric) | 23.9 cm |
| Width (Metric) | 22.7 cm |
| Height (Metric) | 36 cm |
| Holds | Wipes |
| Material | Plastic |
| Mounting | Wall Mount |
| No. of Compartments | 1 |
| Show More |