missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAY 55-9837

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Bay 55-9837 is a selective VPAC2 receptor agonist (EC50 values are 0.4, 100 and >1000 nM for VPAC2, VPAC1 and PAC1, respectively in a cAMP accumulation assay; IC50 values are 60, 8700 and >10000 nM for VPAC2, VPAC1 and PAC1, respectively in a competition binding assay). Stimulates glucose-dependent insulin secretion in isolated human pancreatic islets. Reduces HIV-1 viral replication and shows cooperative effects when given in conjunction with VPAC1 agonists.
Specifications
Specifications
| Purity | 95% |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | M.W. (Observed): 3742.29 |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction