Learn More
MedChemExpress Tocilizumab
Specifications
Specifications
| Antigen | Interleukin Related |
| Quantity | 5 mg |
| Target Species | Human |
| Content And Storage | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Product Type | Inhibitory Antibodies |
| Shipping Condition | Shipping with dry ice |
| CAS | 375823-41-9 |
| Purity or Quality Grade | 98.0% |