missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM87A Antibody (772807) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7966J
This item is not returnable.
View return policy
Description
TMEM87A Monoclonal antibody specifically detects TMEM87A in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF
Specifications
| TMEM87A | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 25963 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Simple Western, Flow Cytometry, CyTOF | |
| 772807 | |
| Western Blot, Simple Western, Flow Cytometry, CyTOF-ready | |
| DKFZp564G2022, PSEC0094, transmembrane protein 87A | |
| E. coli-derived recombinant human TMEM87A, Ser22-Asn157, Accession # Q8NBN3 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction