missing translation for 'onlineSavingsMsg'
Learn More

TMEM74 Mouse Monoclonal Antibody, Proteintech

Product Code. p-200107955 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30233454 150 μL 150µL
30236716 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30233454 Supplier Proteintech Supplier No. 677731IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TMEM74 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMEM74. This antibody recognizes human antigen. The TMEM74 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen TMEM74
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2H1B7
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols FLJ30668
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein G purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157753
Target Species Human
Content And Storage Store at -20°C.
Antibody Molecular Weight 33 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.