missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM41A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20768-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| TMEM41A | |
| Polyclonal | |
| Unconjugated | |
| TMEM41A | |
| 2900010K02Rik, TMEM41A, transmembrane protein 41A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66664, 681708, 90407 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96HV5, Q9D8U2 | |
| TMEM41A | |
| TMEM41A Fusion Protein Ag14753 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction