missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: RWSLVLSPRL EPSGVISAHC NLHLLASSDS SASASRLCQR VMMPSRTNLA TGIPSSKVKY SRLSSTDDGY IDLQFKKTPP KIPYK Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 51%.
Involved in trafficking and recycling of synaptic vesicles.
Specifications
Specifications
| Antigen | TMEM230 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TMEM230 |
| Gene Accession No. | Q96A57 |
| Gene Alias | 5730494N06Rik; AA407821; C20orf30; dJ1116H23.2.1; HSPC274; HSPC274 protein; RGD1307399; TMEM230; Transmembrane protein 230; UNQ2432/PRO4992; UPF0414 transmembrane protein C20orf30; UPF0414 transmembrane protein C20orf30 homolog |
| Gene Symbols | TMEM230 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?