missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TMEM176B (also known as LR8) is a transmembrane protein belongs to the MS4A family of proteins. It may play a role in the process of maturation of dendritic cells. TMEM176B was first discovered in human lung fibroblasts and is associated with human small cell lung carcinoma. Studies suggest that TMEM176B might be involved in cancer and might serve as targets for antiangiogenic therapy of cancer patients. There are two isoforms produced by alternative splicing.
Specifications
Specifications
| Antigen | TMEM176B |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TMEM176B |
| Gene Accession No. | Q3YBM2, Q9R1Q6 |
| Gene Alias | LR8, Protein LR8, TMEM176B, transmembrane protein 176B |
| Gene Symbols | TMEM176B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?