missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM100 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 EUR - 410.00 EUR
Specifications
| Antigen | TMEM100 |
|---|---|
| Concentration | 467 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16864135
|
Proteintech
25581-1-AP-20UL |
20 μL |
145.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16854135
|
Proteintech
25581-1-AP-150UL |
150 μL |
410.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TMEM100 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TMEM100. This antibody recognizes human, mouse antigen. The TMEM100 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| TMEM100 | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 467 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 55273 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title