Learn More
Macherey-Nagel™ TLC Simultaneous Developing Chamber
Product Code. p-8012435 Shop All Macherey-Nagel Products
Click to view available options
Holds:
2
5
Unit Size:
1 set
Each
Product Code. 12374613
Brand: Macherey-Nagel™ 814018
Only null left
Add to basket
Add to basket
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
Develop multiple TLC plates simultaneously. TLC Simultaneous Developing Chambers come with a choice of room for either 2 or 5 plates.
  • Accessory for TLC
  • Chambers are thick walled
  • Clear glass tank 
  • Heavy ground glass lid
Specifications
Quantity 1 Pk.
Type TLC Simultaneous Developing Chamber
For Use With (Application) For 2 Plates 10 x 10 cm
Holds 2