Learn More
BRAND™ Titrette™ Dispensing Cylinder with Valve Block
Product Code. 11653219 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
Titrette 25 mL, from serial-number 01K
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11653219 Titrette 25 mL, from serial-number 01K Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11653219 Supplier BRAND™ Supplier No. 707535
Only null left
Add to basket
Add to basket
Structural change from serial-number 01K
The assembly of the titrating tube to the valve block has been changed. When ordering spare parts, the serial number must be accounted for.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Titrette 25 mL, from serial-number 01K
Includes Valve block